Dionex™ ICS-Series VWD Variable Wavelength Detector Replacement Lamps
Actual product may vary
Thermo Scientific™
Dionex™ ICS-Series VWD Variable Wavelength Detector Replacement Lamps
High-quality Thermo Scientific™ Variable Wavelength Detector Accessories are used for operation of ICS Series Varibale Wavelength Detectors.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberdimType
066349
066348
066347
Catalog number 066349
Price (USD)
-
High-quality Thermo Scientific™ Variable Wavelength Detector Accessories are used for operation of ICS Series Varibale Wavelength Detectors.
Accessories for ICS Series Variable Wavelength Detector
  • Flow cells: Analytical (PEEK), semi-micro (stainless steel)
  • Lamps: Tungsten and deuterium for low noice and high intensity over the full spectral range
  • Digital/analog converter (with cables)
Specifications
DescriptionVWD Option (includes 2 Signal Cables) - Required for output of an Analog signal
Detector TypeVariable Wavelength
For Use With (Application)Liquid Chromatography
For Use With (Equipment)ICS Series Varibale Wavelength Detectors
Product LineThermo Scientific
Unit SizeEach