Dionex™ ASE™ 150/350 Stainless Steel Extraction Cells
Actual product may vary
Dionex™ ASE™ 150/350 Stainless Steel Extraction Cells
Dionex™ ASE™ 150/350 Stainless Steel Extraction Cells
Dionex™ ASE™ 150/350 Stainless Steel Extraction Cells
Dionex™ ASE™ 150/350 Stainless Steel Extraction Cells
Dionex™ ASE™ 150/350 Stainless Steel Extraction Cells
Dionex™ ASE™ 150/350 Stainless Steel Extraction Cells
Thermo Scientific™
Dionex™ ASE™ 150/350 Stainless Steel Extraction Cells
These convenient kits include 1mL to 100mL extraction cells with end caps, seals, and O-rings for Thermo Scientific™ Dionex™ ASE™ 150 and 350 Accelerated Solvent Extractors.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)Quantity
068091100 mL1
0680851 mL1
0680865 mL1
06808710 mL1
06808822 mL1
06808934 mL1
06809066 mL1
0680951 mL6/Pk.
0680965 mL6/Pk.
06809710 mL6/Pk.
06809822 mL6/Pk.
06809934 mL6/Pk.
06810066 mL6/Pk.
068101100 mL6/Pk.
Catalog number 068091
Price (USD)
753.00
Each 1
Volume (Metric):
100 mL
Quantity:
1
Price (USD)
753.00
Each 1
These convenient Thermo Scientific™ Dionex™ ASE™ 150 and 350 Stainless Steel Extraction Cell Kits include 1mL to 100mL extraction cells. Complete with end caps, seals, and O-rings, you can order them as individual cells or in packs of six.
Dionex ASE Accelerated Solvent Extractors reduce sample preparation from hours to minutes by performing automated solvent extraction on solid and semisolid sample matrices. Extraction cells feature finger-tight cell caps with compression seal for high-pressure closure and maximum ease of use.
Kit complete with end caps, seals and O-rings.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Description100 mL Stainless Steel Extraction Cell Kit
Includes1 Extraction Cell, End Caps, Seals, O-Rings
MaterialStainless Steel
Quantity1
Volume (Metric)100 mL
For Use With (Equipment)Dionex ASE 150 and 350
TypeExtraction Cell Kit
Unit SizeEach 1