Dionex™ ASE™ 150/350 Dionium™ Extraction Cells
Actual product may vary
Dionex™ ASE™ 150/350 Dionium™ Extraction Cells
Dionex™ ASE™ 150/350 Dionium™ Extraction Cells
Dionex™ ASE™ 150/350 Dionium™ Extraction Cells
Thermo Scientific™
Dionex™ ASE™ 150/350 Dionium™ Extraction Cells
These Thermo Scientific™ Dionex™ ASE™ 150/350 Dionium™ Extraction Cells and caps are resistant to low concentrations of mineral acids and strong bases used in Dionex ASE 150 and 350 systems.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)Quantity
06810266 mL1
068103100 mL1
06810466 mL3/Pk.
068105100 mL3/Pk.
Catalog number 068102
Price (USD)
1,910.00
Each 1
Volume (Metric):
66 mL
Quantity:
1
Price (USD)
1,910.00
Each 1
Resistant to low concentrations of mineral acids and strong bases, these kits include 66mL or 100mL Dionium extraction cells with end caps, seals and O-ring. Use these Thermo Scientific™ Dionex™ ASE™ 150/350 Dionium™ Extraction Cells with Dionex ASE 150 and 350 Accelerated Solvent Extractors to perform solvent extraction on solid and semisolid samples. The chemically inert Dionium is ideal for samples pretreated with acids or bases. Dionium supports expanded choices of both pretreated matrices and post-treatment adsorbents for in-cell cleanup. Order single extraction cells or a pack of three.
Dionex ASE systems reduce sample preparation from hours to minutes by performing automated solvent extraction on solid and semisolid sample matrices.
Kit complete with end caps, seals and O-rings.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Description66 mL Dionium Extraction Cell Kit
Includes1 Extraction Cell, End Caps, Seals, and O-Rings
MaterialDionium
Quantity1
Volume (Metric)66 mL
For Use With (Equipment)Dionex ASE 150 and 350
TypeExtraction Cell Kit
Unit SizeEach 1