Selenium sulfide, 97%
Selenium sulfide, 97%
Selenium sulfide, 97%
Selenium sulfide, 97%
Thermo Scientific Chemicals

Selenium sulfide, 97%

CAS: 7488-56-4 | S2Se | 143.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
50 g
250 g
Catalog number 068104.06
also known as 068104-06
Price (USD)
41.20
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
41.20
Each
Chemical Identifiers
CAS7488-56-4
IUPAC Namethione
Molecular FormulaS2Se
InChI KeyJNMWHTHYDQTDQZ-UHFFFAOYSA-N
SMILESS=[Se]=S
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)96.1% min
Selenium sulfide is used as an antifungal agent to treat dandruff, seborrheic dermatitis, eborrhea and tinea versicolor. It is the active component in shampoos as an antifungal agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only