Dimethyl selenide
Dimethyl selenide
Dimethyl selenide
Thermo Scientific Chemicals

Dimethyl selenide

CAS: 593-79-3 | C2H6Se | 109.041 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number 068116.14
also known as 068116-14
Price (USD)
768.65
Online Exclusive
854.00
Save 85.35 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
768.65
Online Exclusive
854.00
Save 85.35 (10%)
Each
Chemical Identifiers
CAS593-79-3
IUPAC Name(methylselanyl)methane
Molecular FormulaC2H6Se
InChI KeyRVIXKDRPFPUUOO-UHFFFAOYSA-N
SMILESC[Se]C
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)> 97.0%
FormClear Liquid
Used as primary and secondary intermediate and also used in chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only