Thermo Scientific Chemicals

Ammonium hexafluorosilicate, 98%, Thermo Scientific Chemicals

Catalog number: 069100.30
250 g, Each
Thermo Scientific Chemicals

Ammonium hexafluorosilicate, 98%, Thermo Scientific Chemicals

Catalog number: 069100.30
250 g, Each
Quantity
Catalog number: 069100.30
also known as 069100-30
Price (USD)
Price: 47.00
Online price: 39.65
Your price:
Quantity
-

Chemical Identifiers

CAS
16919-19-0
IUPAC Name
diammonium hexafluorosilanediuide
Molecular Formula
F6H8N2Si
InChI Key
ITHIMUMYFVCXSL-UHFFFAOYSA-P
SMILES
[NH4+].[NH4+].F[Si--](F)(F)(F)(F)F
Assay (unspecified)
97.6% min

Description

Etching glass, in soldering fluxAmmonium hexafluorosilicate is used as a disinfectant and analytical reagent. It finds application in etching glass, metal casting, and electroplating. It is also employed as a wood preservative agent and mothproofing agent in textiles. It also finds application in soldering flux. It is also used to arrest caries treatment without discoloration on demineralized primary tooth enamel.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text