Product Image
Thermo Scientific™

Dionex™ Monobromoacetic Acid-1-13C Internal Standard, 1000 mg/L, 1 mL

Simplify the challenge of determining haloacetic acids (HAA) in drinking water with these convenient, pre-prepared standards IC analysis.
Have Questions?
Catalog NumberDescription
069407

Dionex™ Monobromoacetic Acid-1-13C Internal Standard, 1000 mg/L, 1 mL

Catalog number 069407
Price (USD)
517.00
Each
Description:

Dionex™ Monobromoacetic Acid-1-13C Internal Standard, 1000 mg/L, 1 mL

Price (USD)
517.00
Each
Simplify the challenge of determining haloacetic acids (HAA) in drinking water with these convenient, pre-prepared standards IC analysis to meet U.S. EPA required minimum contamination levels. Designed to save time and eliminate errors in standards preparation, you can rely on Thermo Scientific™ Dionex™ Haloacetic Acid Internal Standards for purity and quality.
Three 1mL HAA internal standards with verified concentrations:
  • Monochloroacetic acid MCAA-2-13C, 1000mg/L
  • Monobromoacetic acid MBAA-1-13C, 1000mg/L
  • Trichloroacetic Acid TCAA-2-13C, 1000mg/L
Quality to Trust
All Thermo Scientific Dionex IC standards are traceable to National Institute of Standards and Technology (NIST™) Standard Reference Materials and are shipped with a Certificate of Analysis verifying the concentration.
Specifications
Description

Dionex™ Monobromoacetic Acid-1-13C Internal Standard, 1000 mg/L, 1 mL

For Use With (Application)ESI/MS Detection
HazardsFlammable, Irritant
OperationUsed as Internal Standard in MtBE
QuantityEach
Standard ComponentsBromoacetic Acid, Methyl t-butyl Ether
Standard Concentration1000 mg/L
Storage<10°C
Volume (Metric)1 mL
TypeInternal Standard
Unit SizeEach