Dionex™ AS-AP Autosampler Trays
Actual product may vary
Thermo Scientific™
Dionex™ AS-AP Autosampler Trays
Thermo Scientific™ Dionex™ AS-AP Autosampler Trays are available to support vials and deep well plates.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescription
069877AS-AP 8-position 10 mL Vial Tray
074936AS-AP 1.5/0.3 mL Vial Tray
074938AS-AP 10 mL Vial Tray
074939AS-AP Support Tray for Deep Well Plates
Catalog number 069877
Price (MXN)
-
Description:
AS-AP 8-position 10 mL Vial Tray
Thermo Scientific™ Dionex™ AS-AP Autosampler Trays are available to support vials and deep well plates.
Specifications
DescriptionAS-AP 8-position 10 mL Vial Tray
For Use With (Equipment)AS-AP Autosampler
Product Type8 Position Vial Tray
Sample Capacity8 Position
Volume (Metric)10 mL Vials
Unit SizeEach