Tin(IV) bromide, 99%
Tin(IV) bromide, 99%
Tin(IV) bromide, 99%
Thermo Scientific Chemicals

Tin(IV) bromide, 99%

CAS: 7789-67-5 | Br4Sn | 438.326 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Catalog number 071120.30
also known as 071120-30
Price (USD)
499.65
Online Exclusive
555.00
Save 55.35 (10%)
Each
Quantity:
250 g
Request bulk or custom format
Price (USD)
499.65
Online Exclusive
555.00
Save 55.35 (10%)
Each
Chemical Identifiers
CAS7789-67-5
IUPAC Nametin(IV) bromide
Molecular FormulaBr4H4Sn
InChI KeyQENMVORAJVNRHX-UHFFFAOYSA-N
SMILESBr.Br.Br.Br.[Sn]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)98.6% min
A specialty product for proteomics research. Used as a precursor for SnO2 through laser-assisted CVD with higher efficiency than other precursors such as dibutyl tin diacetate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only