n-Butyltin trichloride, 96%
n-Butyltin trichloride, 96%
n-Butyltin trichloride, 96%
Thermo Scientific Chemicals

n-Butyltin trichloride, 96%

CAS: 1118-46-3 | C4H9Cl3Sn | 282.176 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 071125.22
also known as 071125-22
Price (USD)
-
Quantity:
100 g
Request bulk or custom format
Specifications
CAS1118-46-3
Chemical Name or Materialn-Butyltin trichloride
DOT InformationTransport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
Density1.693 g/mL
EINECS Number214-263-6
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only