Dimethyltin dichloride
Dimethyltin dichloride
Dimethyltin dichloride
Thermo Scientific Chemicals

Dimethyltin dichloride

CAS: 753-73-1 | C2H6Cl2Sn | 219.68 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2 g
50 g
Catalog number 071142.18
also known as 071142-18
Price (USD)
278.65
Online Exclusive
309.00
Save 30.35 (10%)
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
278.65
Online Exclusive
309.00
Save 30.35 (10%)
Each
Chemical Identifiers
CAS753-73-1
Molecular FormulaC2H8Cl2Sn
InChI KeyQLQTYMCOTSXVHH-UHFFFAOYSA-N
SMILESCl.Cl.C[SnH2]C
Molecular Weight (g/mol)221.70
SpecificationsSpecification SheetSpecification Sheet
Assay from Supplier's CofA≥98.5%
Appearance (Color)White to orange or green
Melting Point (clear melt)106-110°C
FormCrystals or powder or crystalline powder
Used as a pharmaceutical intermediate. It is employed as a coating, plastic and rubber auxilliary agent. Used as methyl Tin stabilizer intermediates, catalysts for esterification reaction, raw material for glass coating.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only