Tetramethyltin, 98%
Tetramethyltin, 98%
Tetramethyltin, 98%
Thermo Scientific Chemicals

Tetramethyltin, 98%

CAS: 594-27-4 | C4H12Sn | 178.85 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number 071145.18
also known as 071145-18
Price (USD)
256.65
Online Exclusive
285.00
Save 28.35 (10%)
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
256.65
Online Exclusive
285.00
Save 28.35 (10%)
Each
Chemical Identifiers
CAS594-27-4
IUPAC Nametetramethylstannane
Molecular FormulaC4H12Sn
InChI KeyVXKWYPOMXBVZSJ-UHFFFAOYSA-N
SMILESC[Sn](C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Refractive Index1.4400-1.4420 @ 20°C (non-U.S. specification)
Assay (GC)≥97.5%
Assay from Suppliers CofA≥97.5% (U.S. specification)
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only