Methyltin trichloride, 97%
Methyltin trichloride, 97%
Methyltin trichloride, 97%
Thermo Scientific Chemicals

Methyltin trichloride, 97%

CAS: 993-16-8 | CH3Cl3Sn | 240.095 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number 071156.14
also known as 071156-14
Price (USD)
1,143.65
Online Exclusive
1,271.00
Save 127.35 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
1,143.65
Online Exclusive
1,271.00
Save 127.35 (10%)
Each
Chemical Identifiers
CAS993-16-8
Molecular FormulaCH9Cl3Sn
InChI KeyVLOZTKJSAQVQRJ-UHFFFAOYSA-N
SMILESCl.Cl.Cl.C[SnH3]
Molecular Weight (g/mol)246.14
SpecificationsSpecification SheetSpecification Sheet
FormPowder or crystals or crystalline powder
Assay from Suppliers CofA≥96.0%
Appearance (Color)White
A chemical used in the synthesis of metallasilesesquiloxanes. Also used in the preparation of organotin(IV) complexes showing antibacterial activities.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only