Phenyltin trichloride
Phenyltin trichloride
Phenyltin trichloride
Thermo Scientific Chemicals

Phenyltin trichloride

CAS: 1124-19-2 | C6H5Cl3Sn | 302.166 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number 071157.14
also known as 071157-14
Price (USD)
303.00
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
303.00
Each
Specifications
CAS1124-19-2
Chemical Name or MaterialPhenyltin trichloride
DOT InformationHazard Class: 8; Packaging Group: II
Density1.84 g/mL
EINECS Number214-393-3
View more
Phenyltin trichloride is used to produce 3-phenyl-cyclohexanone with cyclohex-2-enone. And it is also used in pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only