Tetraallyltin, 96%
Tetraallyltin, 96%
Tetraallyltin, 96%
Thermo Scientific Chemicals

Tetraallyltin, 96%

CAS: 7393-43-3 | C12H20Sn | 283.00 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2 g
10 g
Catalog number 071158.09
also known as 071158-09
Price (USD)
238.65
Special offer
Online exclusive
Ends: 30-Jun-2026
281.00
Save 42.35 (15%)
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
238.65
Special offer
Online exclusive
Ends: 30-Jun-2026
281.00
Save 42.35 (15%)
Each
Specifications
CAS7393-43-3
Chemical Name or MaterialTetraallyltin
DOT InformationTransport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ORGANOTIN COMPOUNDS, LIQUID, N.O.S.
Density1.179 g/mL
EINECS Number230-987-5
View more
Applied as allyl transfer reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only