Trimethyltin chloride
Trimethyltin chloride
Trimethyltin chloride
Thermo Scientific Chemicals

Trimethyltin chloride

CAS: 1066-45-1 | C3H11ClSn | 201.28 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2 g
10 g
50 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 071166.18
also known as 071166-18
Price (USD)
-
Quantity:
50 g
Request bulk or custom format
Chemical Identifiers
CAS1066-45-1
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormCrystals or powder or crystalline powder
Assay (GC)≥98+%
Identification (FTIR)Conforms
Melting Point37-41°C
Trimethyltin chloride is used as a heat stabilizer in the manufacture of vinyl window blinds. It is also used as a stabilizer for plastics and as additive to paint, as antifouling agent. It also serves as a byproduct of methyltin manufacture. Further, it is used as a precursor to vinyltrimethylstannane and in polyvinyl chloride stabilization.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only