Trimethyltin hydroxide, 98%
Trimethyltin hydroxide, 98%
Trimethyltin hydroxide, 98%
Thermo Scientific Chemicals

Trimethyltin hydroxide, 98%

Trimethyltin hydroxide, 98%, C3H11OSn, CAS Number-56-24-6, hydroxytrimethyltin, trimethylstannyl hydroxide, trimethyl tin hydroxide, stannane, hydroxytrimethyl, tin, trimethyl-, hydroxide, trimethyltin hydroxide, hydroxytrimethylstannane, trimethylstannanol, trimethylhydroxytin, unii-10alt8dj2v, 25g | CAS: 56-24-6 | C3H10OSn | 180.82 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 071167.03
also known as 071167-03
Price (USD)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS56-24-6
IUPAC Nametrimethylstannanol
Molecular FormulaC3H10OSn
InChI KeyOJZNYUDKNVNEMV-UHFFFAOYSA-M
SMILESC[Sn](C)(C)O
View more
SpecificationsSpecification SheetSpecification Sheet
FormSolid
Appearance (Color)White to off-white
Assay from Suppliers CofA≥97.5%
Trimethyltin hydroxide is used in the synthesis of modified thymines, which is useful as inhibitors of RNase A. It is also used in the preparation of ketoheterocycle inhibitors of fatty acid amide inhibitors. It reacts with 2,2-disubstituted benzothiazolines to form trialkyltin derivatives of sulphur containing Schiff base complexes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only