Di-n-octyltin oxide
Di-n-octyltin oxide
Di-n-octyltin oxide
Thermo Scientific Chemicals

Di-n-octyltin oxide

CAS: 870-08-6 | C16H34OSn | 361.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 071193.36
also known as 071193-36
Price (USD)
-
Quantity:
500 g
Request bulk or custom format
Specifications
CAS870-08-6
Chemical Name or MaterialDi-n-octyltin oxide
ColorWhite
EINECS Number212-791-1
Formula Weight361.14
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only