Dionex™ ICS-6000 CD Conductivity Detector
Dionex™ ICS-6000 CD Conductivity Detector
Thermo Scientific™
Dionex™ ICS-6000 CD Conductivity Detector
Get high-performance conductivity detection for analytical and capillary ion chromatography separations.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberType
072041Conductivity Detector (Capillary)
063725Remote Housing
079829Conductivity Detector (Analytical)
Catalog number 072041
Price (USD)
-
Request A Quote
Type:
Conductivity Detector (Capillary)
Be ready for any ion chromatography (IC) application, from single column methods in high-background matrices to trace contaminant determinations in high-purity water. The Thermo Scientific™ Dionex™ ICS-6000 CD Conductivity Detector handles even very high or low concentrations with high sensitivity, low noise and without cumbersome range change distortions.
High Sensitivity with Low Noise Across Concentrations:
  • Excellent detection from high to low concentrations with microprocessor-controlled digital signal processing and wide dynamic range of up to 15,000μS/cm
  • Improved stability and lower noise from the integrated electronics between the cell and detector
  • Enhanced thermal stability with independent detector temperature control
  • Choice of capillary and analytical (standard and microbore) models
  • Flexible configuration for use in series with another detector or parallel for dual detection
  • Simple, snap-in installation (no cables or tools required) automatically recognized by Thermo Scientific™ Chromeleon™ Chromatography Data System software
  • Compact installation requiring minimal tubing lengths with up to two detector modules in the same ICS-5000+/ICS-6000 DC Detector/Chromatography Compartment
Capillary CD Detector:
  • Flow- and volume-optimized for capillary applications
  • Easy on sample with minimum cell volume of just 0.02μL
  • No heat exchanger required
Specifications
Automated Start-Up FunctionsStandard
Cell BodyChemically inert polymeric material
Cell ElectrodesPassivated 316 Stainless Steel
Cell Temperature5°C
Cell Temperature Stability<0.00°C
Data Collection Rate1 to 100 Hz
DescriptionConductivity Detector (Capillary)
Electronic Noise<50 Pc.
Electronics TypeMicroprocessor controlled digital signal processing autoranging
Flow Cell Max. Pressure1500 psi
Flow Cell Volume0.02 μL
GLP FeaturesYes
Linearity1%
Output Range0 to 18000 μS/cm
Weight (Metric)400 g
Detector TypeConductivity (CD)
For Use With (Equipment)Dionex ICS-5000 Ion Chromatography System
TypeConductivity Detector (Capillary)
Unit SizeEach