Strontium bromide hexahydrate, 95%
Strontium bromide hexahydrate, 95%
Strontium bromide hexahydrate, 95%
Thermo Scientific Chemicals

Strontium bromide hexahydrate, 95%

CAS: 7789-53-9 | Br2H12O6Sr | 355.518 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number 072105.22
also known as 072105-22
Price (USD)
45.10
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
45.10
Each
Chemical Identifiers
CAS7789-53-9
IUPAC Namestrontium(2+) hexahydrate dibromide
Molecular FormulaBr2H12O6Sr
InChI KeyFLMJUJXBFKFYOZ-UHFFFAOYSA-L
SMILESO.O.O.O.O.O.[Br-].[Br-].[Sr++]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)94.1% min
Strontium bromide hexahydrate is used in flares and also used in some pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only