Dionex™ EO Eluent Organizer Regulator Accessories
Actual product may vary
Thermo Scientific™
Dionex™ EO Eluent Organizer Regulator Accessories
use EO Eluent Organizer Regulator Accessories are for ion chromatography eluent organizers.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTypeFor Use With (Application)
074422EO Regulator AccessoryMounting on the DC Detector
074424EO Regulator Accessory and StandMounting on the TC/VWD/PDA
074423EO Regulator Accessory and StandMounting on the DC Detector
Catalog number 074422
Price (USD)
1,160.00
Each 1
Type:
EO Regulator Accessory
For Use With (Application):
Mounting on the DC Detector
Price (USD)
1,160.00
Each 1
Use Thermo Scientific™ Dionex™ EO Eluent Organizer Regulator Accessories are for ion chromatography eluent organizers. Order the appropriate stand to fit your detector model.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
DescriptionEO Regulator Accessory (to add to 074423 or 074424 for gas regulation of second channel)
For Use With (Application)Mounting on the DC Detector
For Use With (Equipment)EO Regulator Accessories and Stands for Gas Regulation of Second Channel
IncludesEO Regulator kit
OperationProvides the option for gas regulation of the second channel in the Dionex EO Eluent Organizer.
QuantityEach 1
TypeEO Regulator Accessory
Unit SizeEach 1