MAbPac™ SEC-1 Size Exclusion Chromatography HPLC Columns
Thermo Scientific™

MAbPac™ SEC-1 Size Exclusion Chromatography HPLC Columns

Green features

Use these SEC columns for monoclonal antibody (mAb) analysis, including the high-resolution separation of monomers, aggregates and fragments.

Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (Metric)Diameter (Metric)
074696300 mm4 mm
088460300 mm7.8 mm
07469750 mm4 mm
075592150 mm4 mm
088790
also known as 88790
150 mm2.1 mm
088789300 mm2.1 mm
Catalog number 074696
Price (USD)
2,309.21
Each
Length (Metric):
300 mm
Diameter (Metric):
4 mm
Price (USD)
2,309.21
Each

Use Thermo Scientific™ MAbPac™ SEC-1 Size Exclusion Chromatography HPLC Columns for high-resolution separation of monoclonal antibody (mAb) analysis, including the monomers, aggregates and Fab and Fc fragments resulting from proteolysis. These size-exclusion chromatography (SEC) columns provide superior performance, even under nondenaturing conditions, and in high- and low-salt mobile phases and volatile eluents.

High-Performance SEC Columns for mAb Analysis
  • Hydrophilic bonded layer minimizes undesired interactions between the biomolecules and the stationary phase
  • Nonmetallic and biocompatible PEEK housing eliminates metal contamination from the column hardware
  • Stable surface bonding leads to low column bleed and compatibility with MS, ELSD and Thermo Scientific™ Dionex™ Corona™ CAD detection
  • Rugged column packing provides reliability, reproducibility and long column life
  • Columns are available in 5μm particle size and 150 and 300mm lengths, with an average pore diameter of 300Å
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Column ChemistryProprietary Diol
Column FormatAnalytical Column
Column TypeSize Exclusion
Green FeaturesACT Ecolabel (Silver)
Max. Pressure1000 psi (70 bar)
Packing MaterialSpherical, Fully Porous, Ultrapure Silica
QuantityEach
Solvents100% Organic Solvents
Stationary PhaseDiol (OH)
Temperature30°C
USP TypeL59
Diameter (Metric)4 mm
Length (Metric)300 mm
MaterialPEEK
Particle Size5 μm
PhaseSize Exclusion
Pore Size300 Å
Product LineMAbPac
TechniqueBio-LC
pH2.5 to 7.5
Unit SizeEach