Tellurium(IV) bromide, 99.9% (metals basis)
Tellurium(IV) bromide, 99.9% (metals basis)
Tellurium(IV) bromide, 99.9% (metals basis)
Thermo Scientific Chemicals

Tellurium(IV) bromide, 99.9% (metals basis)

CAS: 10031-27-3 | Br4Te | 447.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number 075101.09
also known as 075101-09
Price (USD)
116.65
Online Exclusive
129.00
Save 12.35 (10%)
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
116.65
Online Exclusive
129.00
Save 12.35 (10%)
Each
Chemical Identifiers
CAS10031-27-3
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.1% max.
Used as organic intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only