Acclaim™ C30 HPLC Columns
Thermo Scientific™

Acclaim™ C30 HPLC Columns

Green features
Use for superior separation of hydrophobic structural isomers.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (Metric)Diameter (Metric)Particle Size
075719150 mm4.6 mm5 μm
075718250 mm4.6 mm5 μm
078660100 mm4.6 mm3 μm
07866150 mm4.6 mm3 μm
078664250 mm2.1 mm3 μm
078665100 mm2.1 mm3 μm
07866650 mm2.1 mm3 μm
07572010 mm4.6 mm5 μm
07572110 mm3 mm5 μm
07572210 mm2.1 mm5 μm
075723150 mm4.6 mm3 μm
075724150 mm3 mm3 μm
075725150 mm2.1 mm3 μm
075726250 mm3 mm3 μm
303056250 mm4.6 mm3 μm
Catalog number 075719
Price (USD)
1,038.96
Each
Length (Metric):
150 mm
Diameter (Metric):
4.6 mm
Particle Size:
5 μm
Price (USD)
1,038.96
Each

For separations of hydrophobic, structurally related isomers, Thermo Scientific™ Acclaim™ C30 LC reversed-phase columns provide superior results. The covalent modification of C30 alkyl silane on ultrapure, spherical, porous silica gel gives these columns high shape selectivity. They are fully compatible with various aqueous buffers, useful for a broad application range of analytes, and offer more flexibility in method development.

High Shape Selectivity for the Separation of Structurally Related Isomers
  • Unique selectivity complementary to other reversed-phase columns
  • High efficiency and rugged packing
  • Extremely low bleed, fully compatible with MS
  • Stable between pH 2 and 8
  • Available in 3 and 5μm particle sizes and 4.6, 3.0, and 2.1mm diameters, with an average pore diameter of 200Å
About the Acclaim Family of HPLC Columns

Achieve the high standards set by modern HPLC and LC/MS methods with the Acclaim family of reversed-phase columns. These silica-based columns with advanced column bonding and endcapping technologies provide complementary selectivity, high column efficiencies, and ultra-high resolution for a wide variety of applications.

  • Broad spectrum of stationary phases, including conventional C8 and C18 phases, polar embedded phases, and specialty phases including wide-pore, are available to meet your specific application needs
  • Performance tested to ensure high quality and reliability for pharmaceutical, biopharmaceutical, environmental, food, and other industrial chromatographic separations
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Column FormatAnalytical Column
Column TypeReversed Phase
EndcappedProprietary
Flow Rate1 mL/min
Green FeaturesFSC
Max. Pressure8000 psi (550 bar)
Packing MaterialSpherical, Fully Porous, Ultrapure Silica
QuantityEach
Stationary PhaseC30
Surface Area200 m²/g
Temperature60°C
USP TypeL62
Carbon Load13%
Diameter (Metric)4.6 mm
Length (Metric)150 mm
MaterialStainless Steel
Particle Size5 μm
PhaseReversed Phase
Pore Size200 Å
Product LineAcclaim
TechniqueReversed Phase HPLC
pH2 to 8
Unit SizeEach