Dionex™ Carbonate, Methanesulfonic Acid, and Hydroxide EGC Eluent Generator Cartridges
Actual product may vary
Dionex™ Carbonate, Methanesulfonic Acid, and Hydroxide EGC Eluent Generator Cartridges
Dionex™ Carbonate, Methanesulfonic Acid, and Hydroxide EGC Eluent Generator Cartridges
Dionex™ Carbonate, Methanesulfonic Acid, and Hydroxide EGC Eluent Generator Cartridges
Thermo Scientific™

Dionex™ Carbonate, Methanesulfonic Acid, and Hydroxide EGC Eluent Generator Cartridges

Automate eluent generation and prevent baseline shift, increase sensitivity, improve resolution, and more by using Thermo Scientific™ Dionex™ EGC Cartridges.

Have Questions?
Change viewbuttonViewtableView
Catalog NumberTypeProduct Line
075778KOH Eluent Generator CartridgeDionex EGC 500
088453K2CO3 Eluent Generator CartridgeDionex EGC 500
074532KOH Eluent Generator CartridgeDionex EGC III
074533NaOH Eluent Generator CartridgeDionex EGC III
074534LiOH Eluent Generator CartridgeDionex EGC III
074535MSA Eluent Generator CartridgeDionex EGC III
072076KOH Eluent Generator CartridgeDionex EGC
072077MSA Eluent Generator CartridgeDionex EGC
075779MSA Eluent Generator CartridgeDionex EGC 500
302766KOH Eluent Generator CartridgeDionex EGC 400
302767MSA Eluent Generator CartridgeDionex EGC 400
Catalog number 075778
Price (MXN)
-
Type:
KOH Eluent Generator Cartridge
Product Line:
Dionex EGC 500

Achieve reproducible and reliable chromatography separations using eluent generation. Thermo Scientific™ Dionex™ EGC Eluent Generator Cartridges provide consistent, reproducible eluent generation, ensuring confidence in your results. These cartridges eliminate the manual work of eluent preparation and allow you to focus on the science. Eluent generation also removes the need to handle the acids and bases traditionally required for manual IC eluent preparation, saving cost on chemical storage and disposal.

How Dionex Eluent Generator Cartridges improve IC analysis:

  • Obtain outstanding run-to-run reproducibility by eliminating variability from manual eluent preparation
  • Increase confidence in results by eliminating errors from manual eluent preparation
  • Support large application range
  • Increase workflow efficiency by automating eluent preparation
  • Improve operator's safety by reducing exposure to hazardous chemicals
  • Reduce system maintenance since pump is only exposed to deionized water

Product features:

  • Automatic production of high-purity eluents
  • Support broad range of eluent concentrations
  • Support broad range of flow rates
  • Support pressure up to 5,000 psi
  • Enable isocratic as well as gradient separations
  • Dual EGC mode on Thermo Scientific™ Dionex™ ICS-6000 HPIC System specifically designed for complex carbohydrate analysis (Thermo Scientific™ Dionex™ EGC 400 cartridges)
  • Closed system keeping chemicals contained and isolated

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for six (6) months thereafter.

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Specifications
Concentration0.1 to 100 mM
Flow Rate0.01 to 3.00 mL/min.
For Use With (Equipment)Integrion, ICS-5000+, ICS-6000 HPIC Systems
PressureMaximum: 34.5 MPa (5000 psi)
Product LineDionex EGC 500
QuantityEach
Solvents25% Methanol
TypeKOH Eluent Generator Cartridge
Unit SizeEach