Dionex™ IonPac™ AS25 IC Columns
Actual product may vary
Dionex™ IonPac™ AS25 IC Columns
Dionex™ IonPac™ AS25 IC Columns
Dionex™ IonPac™ AS25 IC Columns
Dionex™ IonPac™ AS25 IC Columns
Dionex™ IonPac™ AS25 IC Columns
Dionex™ IonPac™ AS25 IC Columns
Dionex™ IonPac™ AS25 IC Columns
Thermo Scientific™

Dionex™ IonPac™ AS25 IC Columns

Green features
Separate multivalent and polarizable anions including sulfur species in complex matrices in 25 minutes without the use of solvent.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (Metric)Diameter (Metric)Column Type
07601750 mm2 mmGuard Column
076014250 mm4 mmAnalytical Column
076016250 mm2 mmAnalytical Column
07601550 mm4 mmGuard Column
Catalog number 076017
Price (ZAR)
-
Length (Metric):
50 mm
Diameter (Metric):
2 mm
Column Type:
Guard Column
Separate multivalent and polarizable anions in 25 minutes without solvents using the Thermo Scientific™ Dionex IonPac AS25 column. This high-capacity, hydroxide-selective, anion-exchange column is ideal for the determination of sulfur species in complex matrices such as food and beverage samples as well as wastewater effluents from the industrial, chemical, and petrochemical industries. This column delivers fast, simple separation of polarizable anions, including thiosulfate, iodide, thiocyanate, and perchlorate. For the best peak shape of polarizable anions, use the Thermo Scientific™ Dionex™ IonPac™ AS16 column or the Thermo Scientific™ Dionex™ IonPac AS20 column.
Recommended for Fast Separation of Multivalent Anions in Diverse Sample Matrices
  • Municipal wastewater
  • Food processing samples
  • Paper and pulp effluents
  • Refinery waste products
  • Industrial wastewaters
  • Industrial scrubber solutions
  • Geothermal power plant samples
Superior Chromatographic Performance
  • High capacity: 350μeq per column (4x250mm)
  • Ideal for polyvalent anions, including polysulfates, polyphosphates and polycarboxylates
  • Ultralow hydrophobicity allows fast analysis of polarizable anions
  • Low background signal and enhanced analyte sensitivity with the Thermo Scientific™ Dionex™ Anion Self-Regenerating Suppressor (ASRS™ 300) and a Reagent-Free™ (RFIC™) system
  • Compatible with organic solvents to enhance analyte solubility, modify column selectivity, and effectively clean the column
Available in Multiple Formats

The Dionex IonPac AS25 column is available in formats supporting flow rates from 0.25 to 3mL/min. The microbore format requires less eluent consumption, thereby reducing operating costs.

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
For Use With (Application)Anion Exchange
For Use With (Equipment)IonPac AS25 Analytical Column
Green FeaturesFSC
Max. Pressure3000 psi (207 bar)
QuantityEach 1
Solvent CompatibilitypH 0 to 14, 0 to 100% HPLC Solvents
TypeAS25
Column TypeGuard Column
Diameter (Metric)2 mm
FormatMicrobore
Length (Metric)50 mm
Unit SizeEach 1