Dionex™ IonPac™ CS19 IC Columns
Actual product may vary
Dionex™ IonPac™ CS19 IC Columns
Dionex™ IonPac™ CS19 IC Columns
Thermo Scientific™

Dionex™ IonPac™ CS19 IC Columns

Green features
Separate common inorganic cations, small polar amines and polyvalent amines using a high-capacity carboxylate-functionalized cation-exchange column with simple aqueous eluents.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (Metric)Diameter (Metric)Column Type
076028250 mm2 mmAnalytical Column
076026250 mm4 mmAnalytical Column
07602950 mm2 mmGuard Column
07602750 mm4 mmGuard Column
Catalog number 076028
Price (USD)
1,930.00
Each 1
Length (Metric):
250 mm
Diameter (Metric):
2 mm
Column Type:
Analytical Column
Price (USD)
1,930.00
Each 1

Separate polar amines with the Thermo Scientific™ Dionex™ IonPac™ CS19 Cation-Exchange Column. This high-capacity, carboxylate-functionalized column is specifically optimized for the separation of common cations, small polar amines, and moderately hydrophobic and polyvalent cations using simple aqueous eluents and elevated temperatures. The column is ideal for analysis of power plant waters treated with ammonium, morpholine, or ethanolamine and can also be used to analyze chemical additives, chemical process solutions, scrubber solutions, personal care products, and food samples.

Compared to the Thermo Scientific™ Dionex™ IonPac™ CS19 column, this column uses smaller resin particles for more efficient separations resulting in more accurate peak integration and more reliable results.

Analyze Common Cations and Small Polar Amines in Diverse Matrices
  • Simple gradient separation of small polar amines (including alkanolamines and methylamines), and moderately hydrophobic and polyvalent amines (including biogenic amines and alkyl diamines)
  • Optimized for simple acidic gradient separations with minimal baseline shift
  • High capacity facilitates the analysis of widely disparate peak pairs and high ionic strength matrices with less overloading of the column
  • Compatible with moderate amounts of organic solvents, excluding alcohols, to enhance analyte solubility, modify column selectivity, and facilitate effective column clean-up
Multiple Formats Available for Application Flexibility
  • 4x250mm, 2x250mm analytical columns
  • 4x50mm, 2x50mm guard columns
  • Longer 250mm columns facilitate higher resolution separations of more complex sample matrices
  • Smaller i.d. columns provide higher mass sensitivity, eluent and reagent conservation and lower operating costs
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Column Capacity600 μeq
Column ChemistryAcidic
Crosslinking55% DVB
Flow Rate0.25 to 0.375 mL/min.
For Use With (Application)Cation Exchange
For Use With (Equipment)RFIC Systems
Green FeaturesFSC
HydrophobicityMedium
Max. Pressure3000 psi (207 bar)
Packing MaterialEthylvinylbenzene/Divinylbenzene
Particle Size4 μm
QuantityEach 1
Solvent CompatibilityAcidic Eluents (pH 0 to 7), 0 to 10% HPLC Solvents, Alcohols Should be Avoided
Solvents0 to 10% HPLC Solvents, Alcohols Should be Avoided
Stationary PhaseSuper Macroporous Resin
Column TypeAnalytical Column
Diameter (Metric)2 mm
FormatMicrobore
Length (Metric)250 mm
pH0 to 7
Unit SizeEach 1