Titanium(IV) 2-ethylhexyloxide, 95%
Titanium(IV) 2-ethylhexyloxide, 95%
Titanium(IV) 2-ethylhexyloxide, 95%
Thermo Scientific Chemicals

Titanium(IV) 2-ethylhexyloxide, 95%

CAS: 1070-10-6 | C32H68O4Ti | 564.76 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2 kg
Catalog number 077107.A3
also known as 077107-A3
Price (USD)
269.65
Special offer
Online exclusive
Ends: 30-Jun-2026
317.00
Save 47.35 (15%)
Each
Quantity:
2 kg
Request bulk or custom format
Price (USD)
269.65
Special offer
Online exclusive
Ends: 30-Jun-2026
317.00
Save 47.35 (15%)
Each
Chemical Identifiers
CAS1070-10-6
IUPAC Nametitanium(4+) tetrakis(2-ethylhexan-1-olate)
Molecular FormulaC32H68O4Ti
InChI KeyKTXWGMUMDPYXNN-UHFFFAOYNA-N
SMILES[Ti+4].CCCCC(CC)C[O-].CCCCC(CC)C[O-].CCCCC(CC)C[O-].CCCCC(CC)C[O-]
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid or viscous liquid
Color (APHA)≤100
Assay from Suppliers CofATitanium (Ti): 8.0-9.0%
Chloride content≤100ppm (0.10%)
Appearance (Color)Clear yellow to orange
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only