Titanium(IV) isopropoxide, 97+%
Titanium(IV) isopropoxide, 97+%
Titanium(IV) isopropoxide, 97+%
Thermo Scientific Chemicals

Titanium(IV) isopropoxide, 97+%

Catalyzes the regio selective ring opening of 2,3 epoxy alcohols and acids | CAS: 546-68-9 | C12H28O4Ti | 284.219 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
100 g
500 g
4 x 500 g
Catalog number 077115.09
also known as 077115-09
Price (USD)
42.90
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
42.90
Each
Chemical Identifiers
CAS546-68-9
IUPAC Nametitanium(4+) tetrakis(propan-2-olate)
Molecular FormulaC12H28O4Ti
InChI KeyVXUYXOFXAQZZMF-UHFFFAOYSA-N
SMILES[Ti+4].CC(C)[O-].CC(C)[O-].CC(C)[O-].CC(C)[O-]
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Color (APHA)≤25
Assay (unspecified)Ti: 27.50-28.30% (as TiO2)
Chloride content≤100ppm
Specific Gravity0.950-0.960 @ 25?C
View more
Catalyzes the regio selective ring opening of 2,3 epoxy alcohols and acids.Titanium(IV) isopropoxide is used as a precursor for the preparation of titanium and barium-strontium-titanate thin films. It is useful to make porous titanosilicates and potential ion-exchange materials for cleanup of radioactive wastes. It is an active component of sharpless epoxidation as well as involved in the synthesis of chiral epoxides. In Kulinkovich reaction, it is involved as a catalyst in the preparation of cyclopropanes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only