Titanium(III) oxide, 99.8% (metals basis)
Titanium(III) oxide, 99.8% (metals basis)
Titanium(III) oxide, 99.8% (metals basis)
Titanium(III) oxide, 99.8% (metals basis)
Thermo Scientific Chemicals

Titanium(III) oxide, 99.8% (metals basis)

CAS: 1344-54-3 | O3Ti2 | 143.731 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Catalog number 077127.30
also known as 077127-30
Price (USD)
900.65
Online Exclusive
1,000.00
Save 99.35 (10%)
Each
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (USD)
900.65
Online Exclusive
1,000.00
Save 99.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS1344-54-3
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.2% max.
Titanium(III) oxide is used to study the bioacceptability of medical implant surfaces.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only