Titanium(IV) oxide bis(2,4-pentanedionate)
Titanium(IV) oxide bis(2,4-pentanedionate)
Titanium(IV) oxide bis(2,4-pentanedionate)
Thermo Scientific Chemicals

Titanium(IV) oxide bis(2,4-pentanedionate)

CAS: 14024-64-7 | C10H16O5Ti | 264.1 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Catalog number 077130.18
also known as 077130-18
Price (USD)
314.65
Online Exclusive
349.00
Save 34.35 (10%)
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
314.65
Online Exclusive
349.00
Save 34.35 (10%)
Each
Chemical Identifiers
CAS14024-64-7
IUPAC Namebis(4-hydroxypent-3-en-2-one); oxotitanium
Molecular FormulaC10H16O5Ti
InChI KeyADVORQMAWLEPOI-UHFFFAOYSA-N
SMILESO=[Ti].CC(O)=CC(C)=O.CC(O)=CC(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay from Supplier's CofA≥98.0% (titration)
FormCrystals or crystalline powder
Appearance (Color)Pale yellow
Employed as a cross linking agent for cellulose fibers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only