Dionex™ IonPac™ CS19-4μm IC Columns
Actual product may vary
Dionex™ IonPac™ CS19-4μm IC Columns
Dionex™ IonPac™ CS19-4μm IC Columns
Thermo Scientific™

Dionex™ IonPac™ CS19-4μm IC Columns

Green features
Combine speed with high resolution separations of common inorganic cations, small polar amines and polyvalent amines using this high-capacity cation-exchange column.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (Metric)Diameter (Metric)Column Type
078837250 mm4 mmAnalytical Column
078835250 mm0.4 mmCapillary Separator
07884050 mm4 mmGuard Column
078836250 mm2 mmSeparator Column
07883850 mm0.4 mmGuard Column
07883950 mm2 mmGuard Column
Catalog number 078837
Price (USD)
-
Length (Metric):
250 mm
Diameter (Metric):
4 mm
Column Type:
Analytical Column
Attain the optimal combination of chromatographic speed and high resolution using the new Thermo Scientific™ Dionex™ IonPac™ CS19-4μm Capillary, Analytical, & Guard Column. This high-capacity, high-resolution carboxylate-functionalized column provides excellent resolution of common cations, small polar amines, and polyvalent amines in complex sample matrices such as power plant waters treated with ammonium, morpholine, or ethanolamine, chemical additives, chemical process solutions, scrubber solutions, personal care products, and food samples.
Compared to the Thermo Scientific™ Dionex™ IonPac™ CS19 column, this column uses smaller resin particles for more efficient separations resulting in more accurate peak integration and more reliable results.

Highest Resolution Separation of Common Cations and Small Polar Amines in Diverse Matrices
  • Highest resolution, through the use of smaller resin particles, of common cations and small polar amines in complex sample matrices
  • Higher peak efficiency and increased peak resolution for more consistent integration and more reliable results
  • Easily transfer applications from the Dionex IonPac CS19 column to the 4 μm column due to the similar selectivity
  • Simple gradient separation of small polar amines (including alkanolamines and methylamines), and moderately hydrophobic and polyvalent amines (including biogenic amines and alkyl diamines)
  • High capacity facilitates injection of more concentrated samples without column overloading or peak broadening
  • Compatible with moderate amounts of organic solvents, excluding alcohols, to enhance analyte solubility, modify column selectivity, and facilitate effective column clean-up
Use With a High-Pressure IC System
Higher resolution is achieved through the use of smaller resin particles, which produce a column backpressure over 3000 psi at the standard flow rates. A high-pressure IC system, such as the Thermo Scientific™ Dionex™ ICS-5000+ HPIC™ or the Dionex ICS-4000 Capillary HPIC system is required to operate this column under standard conditions and take advantage of elevated flow rates and faster run times.

Reduce Operating Costs With Lower Eluent Requirements
The Dionex IonPac CS19-4μm capillary column provides the same performance as the standard bore and microbore Dionex IonPac CS19-4μm columns because it is packed with the same material. Operating costs are reduced because the capillary system operates at only 1% of the eluent flow rate of the standard bore system, requiring less eluent and producing less waste.

Multiple Formats Available for Application Flexibility

  • 4x250mm, 2x250mm analytical columns
  • 4x50mm, 2x50mm guard columns
  • 0.4x250mm capillary column, 0.4x50mm capillary guard column
  • Longer 250mm columns facilitate higher resolution separations of more complex sample matrices
  • Smaller i.d. columns provide higher mass sensitivity, eluent and reagent conservation and lower operating costs
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Column Capacity2410 μeq
Column Construction

PEEK with 10-32 Threaded Ferrule-Style End Fittings. All Components Are Nonmetallic.

Crosslinking55% DVB
For Use With (Application)Cation Exchange
Green FeaturesFSC
HydrophobicityMedium
Ion Exchange GroupGrafted Carboxylic Acid
Max. Pressure5000 psi (345 bar)
Particle Size4 μm
QuantityEach 1
Solvent CompatibilityAcidic Eluents (pH 0 to 7), 10% HPLC Solvents, Alcohols Should be Avoided
Solvents

10% HPLC Solvents, Alcohols Should be Avoided.

Stationary PhaseIonPac CS19-4μm
Column TypeAnalytical Column
Diameter (Metric)4 mm
FormatStandard Bore
Length (Metric)250 mm
pH

0 to 7

Unit SizeEach 1