Dionex&trade; ICS-5000<sup>+</sup>/ICS-6000 ED Electrochemical Conventional Electrodes
Actual product may vary
Thermo Scientific™
Dionex™ ICS-5000+/ICS-6000 ED Electrochemical Conventional Electrodes
Obtain Gold, AAA Gold, Platinum, Glassy Carbon or Silver ED electrodes with gasket and polishing kit for Dionex ICS-5000+/ICS-6000 ED Electrochemical Conventional Electrodes.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberType
079854Carbon Electrode
063722Gold Electrode (AAA)
079850Gold Electrode
079851Platinum Electrode
079856Silver Electrode
063723Gold Electrode (3 mm)
Catalog number 079854
Price (USD)
1,800.00
Each 1
Type:
Carbon Electrode
Price (USD)
1,800.00
Each 1
Offer a choice of Gold, AAA Gold, Platinum, Glassy Carbon and Silver, each with gasket and polishing kit with Thermo Scientific™ Dionex™ ICS-5000+/ICS-6000 ED Electrochemical Conventional Electrodes.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
For Use With (Equipment)ICS-5000+/ICS-6000 Ion Chromatography Systems
IncludesGasket, polishing kit
MaterialGlassy Carbon
QuantityEach 1
TypeCarbon Electrode
Unit SizeEach 1