B·R·A·H·M·S™ uE3 KRYPTOR™
Thermo Scientific™
B·R·A·H·M·S™ uE3 KRYPTOR™
B·R·A·H·M·S provides a complete portfolio of biomarkers for the most reliable results in prenatal screening. Get the quadruple test for trisomy screening in second trimester of pregnancy with B·R·A·H·M·S uE3 KRYPTOR in less than 40 minutes.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberType
080391NCalibrator
803075NImmunofluorescent Assay
080392NControls
Catalog number 080391N
Price (MXN)
-
Type:
Calibrator
B·R·A·H·M·S provides a complete portfolio of biomarkers for the screening of chromosomal abnormalities in first and second trimester of pregnancy. B·R·A·H·M·S uE3 KRYPTOR can be used as an aid to provide an early identification of the pregnancies at risk of trisomy 21 in second trimester.
US Only: Product is not 510k cleared in US.
Get confidence in your results with the high precision of B·R·A·H·M·S uE3 KRYPTOR and the full quadruple test on B·R·A·H·M·S KRYPTOR GOLD.
  • Incubation time of only 19 minutes for a short turnaround time
  • Allows measurement in serum
  • Large capacity with 75 tests
  • Kit stable up to 15 days after opening onboard the instrument
  • Easy workflow with only 2 calibrators and one calibration every week
  • Fits your throughput need with B·R·A·H·M·S KRYPTOR compact PLUS or B·R·A·H·M·S KRYPTOR GOLD
Thermo Fisher Scientific products are distributed globally and their uses, applications, indications, claims and availability of products in each country depend on local regulatory marketing authorization status, please consult the Instructions For Use (IFU) available in your country.
Specifications
Analysis Time19 minutes
DescriptionCalibrator (2 levels)
For Use With (Application)Readjust the standard curve memorized in B·R·A·H·M·S KRYPTOR compact PLUS/KRYPTOR GOLD
For Use With (Equipment)B·R·A·H·M·S uE3 KRYPTOR
IncludesCalibrator kit, 12 vials
TypeCalibrator
Detectable AnalytesuE3
FormLyophilized
Unit SizeEach
Contents & Storage
Calibrator kit, 12 vials, 2°C to 8°C