B·R·A·H·M·S™ uE3 KRYPTOR™
Thermo Scientific™
B·R·A·H·M·S™ uE3 KRYPTOR™
B·R·A·H·M·S provides a complete portfolio of biomarkers for the most reliable results in prenatal screening. Get the quadruple test for trisomy screening in second trimester of pregnancy with B·R·A·H·M·S uE3 KRYPTOR in less than 40 minutes.
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoTipo
080392NControles
803075NEnsayos de inmunofluorescencia
080391NCalibrador
Número de catálogo 080392N
Precio (MXN)
-
Tipo:
Controles
B·R·A·H·M·S provides a complete portfolio of biomarkers for the screening of chromosomal abnormalities in first and second trimester of pregnancy. B·R·A·H·M·S uE3 KRYPTOR can be used as an aid to provide an early identification of the pregnancies at risk of trisomy 21 in second trimester.
US Only: Product is not 510k cleared in US.
Get confidence in your results with the high precision of B·R·A·H·M·S uE3 KRYPTOR and the full quadruple test on B·R·A·H·M·S KRYPTOR GOLD.
  • Incubation time of only 19 minutes for a short turnaround time
  • Allows measurement in serum
  • Large capacity with 75 tests
  • Kit stable up to 15 days after opening onboard the instrument
  • Easy workflow with only 2 calibrators and one calibration every week
  • Fits your throughput need with B·R·A·H·M·S KRYPTOR compact PLUS or B·R·A·H·M·S KRYPTOR GOLD
Thermo Fisher Scientific products are distributed globally and their uses, applications, indications, claims and availability of products in each country depend on local regulatory marketing authorization status, please consult the Instructions For Use (IFU) available in your country.
Especificaciones
Tiempo de análisis19 minutes
DescripciónControls (3 levels)
Para utilizar con (aplicación)Quality Control for B·R·A·H·M·S uE3 KRYPTOR assay
Para utilizar con (equipo)B·R·A·H·M·S uE3 KRYPTOR
IncluyeControls kit, 6 vials
TipoControles
Analitos detectablesuE3
FormularioLyophilized
Unit SizeEach
Contenido y almacenamiento
Controls kit, 6 vials, 2°C to 8°C