Orion™ Membrane Caps for Polarographic Dissolved Oxygen Probe
Thermo Scientific™
Orion™ Membrane Caps for Polarographic Dissolved Oxygen Probe
Reduce probe servicing time and eliminate assembly problems using the Thermo Scientific™ Orion™ Membrane Caps for Polarographic Dissolved Oxygen Probe.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFor Use With (Equipment)Quantity
081003Orion DO Probes 080510MD, 081010MD, 0810103 Caps
080515Orion DO Probes 083005MD, 083010MD, 083025MD, 083060MD, 086030MD, 083005A, 083010A, 083005D, 086020A, 081010F1 Cap
Catalog number 081003
Price (USD)
160.20
Online exclusive
178.00
Save 17.80
Each
For Use With (Equipment):
Orion DO Probes 080510MD, 081010MD, 081010
Quantity:
3 Caps
Price (USD)
160.20
Online exclusive
178.00
Save 17.80
Each
Reduce probe servicing time and eliminate assembly problems using Thermo Scientific™ Orion™ Membrane Caps for Polarographic Dissolved Oxygen Probe. Convenient screw-on membrane caps help ensure optimal probe performance.
Warranty: Out of box
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Probe TypePolarographic DO
Quantity3 Caps
For Use With (Equipment)Orion DO Probes 080510MD, 081010MD, 081010
TypeMembrane Cap
Unit SizeEach