Orion™ Protective Guard for Conductivity Cells and Dissolved Oxygen Probes, Plastic
Orion™ Protective Guard for Conductivity Cells and Dissolved Oxygen Probes, Plastic
Orion™ Protective Guard for Conductivity Cells and Dissolved Oxygen Probes, Plastic
Orion™ Protective Guard for Conductivity Cells and Dissolved Oxygen Probes, Plastic
Thermo Scientific™
Orion™ Protective Guard for Conductivity Cells and Dissolved Oxygen Probes, Plastic
Provide additional protection and weight with these protective guards for conductivity cells and dissolved oxygen probes.
Have Questions?
Catalog NumberFor Use With (Equipment)
081045Conductivity Cells, DO Probes
Catalog number 081045
Price (USD)
184.50
Online exclusive
205.00
Save 20.50
Each
For Use With (Equipment):
Conductivity Cells, DO Probes
Price (USD)
184.50
Online exclusive
205.00
Save 20.50
Each
Provide additional protection and weight for conductivity cells and DO probes with Thermo Scientific™ Orion™ Protective Guards for Conductivity Cells and Dissolved Oxygen Probes. Use the all stainless-steel guard when optimum protection and weight are required. Combine the stainless steel cap and plastic sleeve when weight for submersion and standard protection is needed. Or, when minimal weight and protection are required, use the all-plastic guard.
For Use With
  • Conductivity Cells: 013005MD, 013010MD, 0130020MD, 013025MD, 013005A, 013010A, and 013005D
  • Dissolved Oxygen Probes: 080510MD, 0830005MD, 083010MD, 083025MD, 083060MD, 083005A, 083010A, and 083005D
Specifications
DescriptionPlastic
MaterialStainless Steel, Plastic
For Use With (Equipment)Conductivity Cells, DO Probes
TypeProtective Guard
Unit SizeEach