Yttrium(III) fluoride, 98%
Yttrium(III) fluoride, 98%
Yttrium(III) fluoride, 98%
Thermo Scientific Chemicals

Yttrium(III) fluoride, 98%

CAS: 13709-49-4 | F3Y | 145.90 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number 083106.14
also known as 083106-14
Price (USD)
58.65
Online Exclusive
64.50
Save 5.85 (9%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
58.65
Online Exclusive
64.50
Save 5.85 (9%)
Each
Chemical Identifiers
CAS13709-49-4
IUPAC Nameyttrium(3+) trifluoride
Molecular FormulaF3Y
InChI KeyRBORBHYCVONNJH-UHFFFAOYSA-K
SMILES[F-].[F-].[F-].[Y+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)97.6% min
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only