Dionex™ CRS™ 500 and ACRS-ICE 500 Chemically Regenerated Suppressors
Actual product may vary
Thermo Scientific™
Dionex™ CRS™ 500 and ACRS-ICE 500 Chemically Regenerated Suppressors
Lower your ion-exchange chromatography detection limits with high-capacity, solvent-compatible chemically regenerated suppression.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescriptionDiameter
084715ACRS-ICE 500 Suppressor9 mm
085090ACRS 500 Suppressor4 mm
085091ACRS 500 Suppressor2 mm
085092CCRS 500 Suppressor4 mm
085093CCRS 500 Suppressor2 mm
084714ACRS-ICE 500 Suppressor4 mm
Catalog number 084715
Price (USD)
1,440.00
Each 1
Description:
ACRS-ICE 500 Suppressor
Diameter:
9 mm
Price (USD)
1,440.00
Each 1
Lower your ion chromatography (IC) detection limits with low-noise using Thermo Scientific™ Dionex™ CRS™ 500 and ACRS-ICE 500 Chemically Regenerated Suppressors. In a choice of 2, 4mm formats, the Dionex CRS 500 suppressor's high capacity and solvent compatible design can handle high complex sample matrices. The Dionex ACRS-ICE 500 suppressor is ideal for food and beverage products, biological samples, fermentation processes, industrial process liquors, and treated wastewaters.
High-Capacity, Solvent-Compatible Suppression for Ion-Exchange Chromatography
  • Very low noise, chemically regenerated suppressor
  • Low method detection limits
  • Excellent baseline stability
  • Fast start-up and equilibration
  • Easy to use, maintenance-free operation with the DCR kit
  • Improved peak efficiency for improved performance, particularly when coupled with 4μm based columns
  • Improved thermal stability to eliminate installation failures due to damage during shipment, handling, and operation
The Dionex ACRS-ICE 500 suppressor is optimized for organic acid and alcohol analysis in complex or high-ionic strength samples. 
Note: For lowest noise and longest life, this suppressor is recommended for anion and cation separations using eluents containing high-performance liquid chromatography (HPLC) solvents.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for twenty-four (24) months thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
DescriptionACRS-ICE 500 Suppressor
Diameter9 mm
For Use With (Application)Ion Chromatography
OperationIon-exclusion chromatography of organic acids and alcohols in complex or high-ionic strength samples.
QuantityEach 1
Void Volume<50 μL
TypeChemically Regenerated Suppressor
Unit SizeEach 1