What are the benefits of CaptureSelect affinity products?
CaptureSelect affinity resins can make the purification of biomolecules that do not have a traditional affinity purification solution much more efficient. When a targeted, specific affinity purification solution does not exist for a biomolecule, the protein purification scheme can be very complex needing 4-5 chromatography steps. Many chromatographic steps need to be utilized to separate the product of interest from key process and product related impurities. As the number of required purification unit operations increases, the product yield decreases and is heavily impacted with each step added. Yield drives cost of goods, so not having an affinity purification solution can greatly impact the cost for biotherapeutic manufacturing. Using in-house capabilities and expertise, CaptureSelect Custom Services design product-specific affinity ligand based on single-chain, camelid-derived antibodies, and couple that ligand to a high-performance, cost-effective affinity resin. We currently offer off-the-shelf bioprocess resins for antibody fragments, biosimilars, and viral vectors. Custom ligands can be used for biomolecule purification, scavenging of challenging impurities, and quantitation or detection of biomolecules. Custom resins are suitable for use in a cGMP production process.