Dionex™ CRS™ 500 and ACRS-ICE 500 Chemically Regenerated Suppressors
Actual product may vary
Thermo Scientific™
Dionex™ CRS™ 500 and ACRS-ICE 500 Chemically Regenerated Suppressors
Lower your ion-exchange chromatography detection limits with high-capacity, solvent-compatible chemically regenerated suppression.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescriptionDiameter
085090ACRS 500 Suppressor4 mm
085091ACRS 500 Suppressor2 mm
085092CCRS 500 Suppressor4 mm
085093CCRS 500 Suppressor2 mm
084714ACRS-ICE 500 Suppressor4 mm
084715ACRS-ICE 500 Suppressor9 mm
Catalog number 085090
Price (USD)
1,590.00
Each
Description:
ACRS 500 Suppressor
Diameter:
4 mm
Price (USD)
1,590.00
Each
Lower your ion chromatography (IC) detection limits with low-noise using Thermo Scientific™ Dionex™ CRS™ 500 and ACRS-ICE 500 Chemically Regenerated Suppressors. In a choice of 2, 4mm formats, the Dionex CRS 500 suppressor's high capacity and solvent compatible design can handle high complex sample matrices. The Dionex ACRS-ICE 500 suppressor is ideal for food and beverage products, biological samples, fermentation processes, industrial process liquors, and treated wastewaters.
High-Capacity, Solvent-Compatible Suppression for Ion-Exchange Chromatography
  • Very low noise, chemically regenerated suppressor
  • Low method detection limits
  • Excellent baseline stability
  • Fast start-up and equilibration
  • Easy to use, maintenance-free operation with the DCR kit
  • Improved peak efficiency for improved performance, particularly when coupled with 4μm based columns
  • Improved thermal stability to eliminate installation failures due to damage during shipment, handling, and operation
The Dionex ACRS-ICE 500 suppressor is optimized for organic acid and alcohol analysis in complex or high-ionic strength samples. 
Note: For lowest noise and longest life, this suppressor is recommended for anion and cation separations using eluents containing high-performance liquid chromatography (HPLC) solvents.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for twenty-four (24) months thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
DescriptionACRS 500 Suppressor
Diameter4 mm
For Use With (Application)Ion Chromatography
OperationChemically regenerated with very low noise and quick startup time.
QuantityEach 1
Void Volume<50 μL
TypeChemically Regenerated Suppressor
Unit SizeEach