B·R·A·H·M·S™ Inhibin A KRYPTOR™
Thermo Scientific™
B·R·A·H·M·S™ Inhibin A KRYPTOR™
B·R·A·H·M·S provides a complete portfolio of biomarkers for the most reliable results in prenatal screening. Get the quadruple test for trisomy screening in second trimester of pregnancy with B·R·A·H·M·S Inhibin A KRYPTOR in less than 40 minutes.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberType
085091NCalibrator
850075NImmunofluorescent Assay
085092NControls
Catalog number 085091N
Price (USD)
-
Type:
Calibrator
B·R·A·H·M·S provides a complete portfolio of biomarkers for the screening of chromosomal abnormalities in first and second trimester of pregnancy. B·R·A·H·M·S Inhibin A KRYPTOR provides an aid in early identification of the pregnancies at risk of trisomy 21 in second trimester.
US Only: Product is not 510k cleared in US.
Get confidence in your results with the high precision of B·R·A·H·M·S Inhibin A KRYPTOR on B·R·A·H·M·S KRYPTOR GOLD and its short incubation time.
  • Incubation time of only 39 minutes for a short turnaround time
  • Allows measurement in serum
  • Large capacity with 75 tests
  • Kit stable up to 15 days after opening onboard the instrument
  • Easy workflow with only 1 calibrator and one calibration every week
  • Fits your throughput need with B·R·A·H·M·S KRYPTOR GOLD
Thermo Fisher Scientific products are distributed globally and their uses, applications, indications, claims and availability of products in each country depend on local regulatory marketing authorization status, please consult the Instructions For Use (IFU) available in your country.
Specifications
Analysis Time39 minutes
DescriptionCalibrator (1 level)
For Use With (Application)Readjust the standard curve memorized in B·R·A·H·M·S KRYPTOR GOLD
For Use With (Equipment)B·R·A·H·M·S Inhibin A KRYPTOR
IncludesCalibrator kit, 6 vials
TypeCalibrator
Detectable AnalytesInhibin A
FormLyophilized
Unit SizeEach
Contents & Storage
Calibrator kit, 6 vials, 2°C to 8°C