B·R·A·H·M·S™ Inhibin A KRYPTOR™
Thermo Scientific™
B·R·A·H·M·S™ Inhibin A KRYPTOR™
B·R·A·H·M·S provides a complete portfolio of biomarkers for the most reliable results in prenatal screening. Get the quadruple test for trisomy screening in second trimester of pregnancy with B·R·A·H·M·S Inhibin A KRYPTOR in less than 40 minutes.
Have Questions?
Ansicht ändernbuttonViewtableView
KatalognummerTyp
085092NKontrollen
850075NImmunofluorescent Assay
085091NKalibrator
Katalognummer 085092N
Preis (EUR)
-
Typ:
Kontrollen
B·R·A·H·M·S provides a complete portfolio of biomarkers for the screening of chromosomal abnormalities in first and second trimester of pregnancy. B·R·A·H·M·S Inhibin A KRYPTOR provides an aid in early identification of the pregnancies at risk of trisomy 21 in second trimester.
US Only: Product is not 510k cleared in US.
Get confidence in your results with the high precision of B·R·A·H·M·S Inhibin A KRYPTOR on B·R·A·H·M·S KRYPTOR GOLD and its short incubation time.
  • Incubation time of only 39 minutes for a short turnaround time
  • Allows measurement in serum
  • Large capacity with 75 tests
  • Kit stable up to 15 days after opening onboard the instrument
  • Easy workflow with only 1 calibrator and one calibration every week
  • Fits your throughput need with B·R·A·H·M·S KRYPTOR GOLD
Thermo Fisher Scientific products are distributed globally and their uses, applications, indications, claims and availability of products in each country depend on local regulatory marketing authorization status, please consult the Instructions For Use (IFU) available in your country.
Specifications
Analysezeit39 minutes
BeschreibungControls (3 levels)
Zur Verwendung mit (Anwendung)Quality Control for B·R·A·H·M·S Inhibin A KRYPTOR assay
Zur Verwendung mit (Geräte)B·R·A·H·M·S Inhibin A KRYPTOR
EnthältControls kit, 6 vials
TypKontrollen
Nachweisbare AnalytenInhibin A
FormLyophilisiert
Unit SizeBox with 6 vials
Inhalt und Lagerung
Controls kit, 6 vials, 2°C to 8°C