Cadmium phosphide, 99.5% (metals basis)
Cadmium phosphide, 99.5% (metals basis)
Thermo Scientific™

Cadmium phosphide, 99.5% (metals basis)

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
10 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 087219.09
also known as 087219-09
Price (USD)
-
Quantity:
10 g
Request bulk or custom format
Chemical Identifiers
CAS12014-28-7
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Assay from Supplier's CofA≥99.5% (metals basis)
Particle Sizeca. 100 Mesh
WARNING: Cancer – www.P65Warnings.ca.gov