Boron triiodide, 95%
Boron triiodide, 95%
Boron triiodide, 95%
Thermo Scientific Chemicals

Boron triiodide, 95%

CAS: 13517-10-7 | BI3 | 391.52 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 087220.06
also known as 087220-06
Price (USD)
474.65
Online Exclusive
527.00
Save 52.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
474.65
Online Exclusive
527.00
Save 52.35 (10%)
Each
Chemical Identifiers
CAS13517-10-7
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)>94.0%
Boron triiodide is used as pharmaceutical, chemical and organic intermediate. It is also used as a catalyst and for preparation of boron compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only