Magnesium nitrate hexahydrate, 99.97% (metals basis)
Magnesium nitrate hexahydrate, 99.97% (metals basis)
Magnesium nitrate hexahydrate, 99.97% (metals basis)
Thermo Scientific Chemicals

Magnesium nitrate hexahydrate, 99.97% (metals basis)

CAS: 13446-18-9 | H12MgN2O12 | 256.40 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
50 g
250 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 087609.06
also known as 087609-06
Price (USD)
38.65
Online exclusive
42.90
Save 4.25
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
38.65
Online exclusive
42.90
Save 4.25
Each
Chemical Identifiers
CAS13446-18-9
IUPAC Namemagnesium(2+) hexahydrate dinitrate
Molecular FormulaH12MgN2O12
InChI KeyMFUVDXOKPBAHMC-UHFFFAOYSA-N
SMILESO.O.O.O.O.O.[Mg++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.03% max.

It is used in printing, chemical, agriculture and the ceramic industry. It is also used in the detection of selenium in plasma samples and concentration of nitric acid. Its fertilizer grade has 10.5% nitrogen and 9.4% magnesium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only