Potassium iodide, 99.99% (metals basis)
Potassium iodide, 99.99% (metals basis)
Potassium iodide, 99.99% (metals basis)
Thermo Scientific Chemicals

Potassium iodide, 99.99% (metals basis)

CAS: 7681-11-0 | IK | 166.003 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number 087627.14
also known as 087627-14
Price (USD)
101.65
Special offer
Online exclusive
Ends: 28-Dec-2026
119.00
Save 17.35
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
101.65
Special offer
Online exclusive
Ends: 28-Dec-2026
119.00
Save 17.35
Each
Chemical Identifiers
CAS7681-11-0
IUPAC Namepotassium iodide
Molecular FormulaIK
InChI KeyNLKNQRATVPKPDG-UHFFFAOYSA-M
SMILES[K+].[I-]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.01% max.
Potassium iodide is a simple iodide salt utilized as a fluorescence quenching agent which acts by collisional quenching of fluorescent substances by the iodide ion. It is also used in the isolation of single stranded DNA.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only