Antimony(III) acetate, 97%
Antimony(III) acetate, 97%
Antimony(III) acetate, 97%
Thermo Scientific Chemicals

Antimony(III) acetate, 97%

CAS: 6923-52-0 | C6H9O6Sb | 298.892 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
100 g
500 g
2 kg
Catalog number 087657.A3
also known as 087657-A3
Price (USD)
603.65
Special offer
Online exclusive
Ends: 30-Jun-2026
710.00
Save 106.35 (15%)
Each
Quantity:
2 kg
Request bulk or custom format
Price (USD)
603.65
Special offer
Online exclusive
Ends: 30-Jun-2026
710.00
Save 106.35 (15%)
Each
Chemical Identifiers
CAS6923-52-0
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
Elemental Analysis% Sb 40 - 42
Trace Metals< 20 ppm Fe
Chloride content< 20 ppm
Formcrystalline powder
Antimony(III) acetate is a catalyst used in the production of synthetic fibers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only