Sodium borohydride, stable aq. soln., 4.4M in 14M NaOH
Sodium borohydride, stable aq. soln., 4.4M in 14M NaOH
Sodium borohydride, stable aq. soln., 4.4M in 14M NaOH
Thermo Scientific Chemicals

Sodium borohydride, stable aq. soln., 4.4M in 14M NaOH

CAS: 16940-66-2 | BH4Na | 37.83 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
2 L
Catalog number 087659.AP
also known as 087659-AP
Price (USD)
72.65
Online Exclusive
80.70
Save 8.05 (10%)
Each
Quantity:
500 mL
Request bulk or custom format
Price (USD)
72.65
Online Exclusive
80.70
Save 8.05 (10%)
Each
Chemical Identifiers
CAS16940-66-2
IUPAC Namesodium boranuide
Molecular FormulaBH4Na
InChI KeyYOQDYZUWIQVZSF-UHFFFAOYSA-N
SMILES[BH4-].[Na+]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)NaBH₄ 11.85û12.15%
Assay (unspecified)NaOH 38.00-42.00%
Sodium borohydride is used as a reagent in the reduction of amino acids and their derivatives. Also used as a catalyst in ammonia borane dehydrogenation. It acts as a reducing agent in the synthesis of gold nanoparticles. It is also used for wood pulp and as a blowing agent for plastics. It reduces aldehyde and keto groups to give the corresponding alcohols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only