Sodium pentacyanonitrosylferrate(III) dihydrate, ACS, 99.0-102.0%
Sodium pentacyanonitrosylferrate(III) dihydrate, ACS, 99.0-102.0%
Sodium pentacyanonitrosylferrate(III) dihydrate, ACS, 99.0-102.0%
Thermo Scientific Chemicals

Sodium pentacyanonitrosylferrate(III) dihydrate, ACS, 99.0-102.0%

For Chromatographic Detection of Peptides | CAS: 13755-38-9 | C5H4FeN6Na2O3 | 297.95 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Catalog number 087683.18
also known as 087683-18
Price (USD)
79.00
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
79.00
Each
Chemical Identifiers
CAS13755-38-9
IUPAC Namedisodium pentacyano(oxidaniumylidyneamino)ironbis(ylium) dihydrate
Molecular FormulaC5H4FeN6Na2O3
InChI KeyJFPDVRGGFBUORC-UHFFFAOYSA-N
SMILESO.O.[Na+].[Na+].N#C[Fe++](C#N)(C#N)(C#N)(C#N)[N]#[O+]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)99.0-102.0% C₅FeN₆Na₂O.2H2O
Impurity contentSO₄ P.T. (limit about 0.01%)
Insoluble material0.01%
Impurity contentCl 0.02%
For chromatographic detection of peptides.Sodium pentacyanonitrosylferrate(III) dihydrate, is used as a vasodilator drug to reduce blood pressure. It is used as an analytical reagent for the detection of methyl ketones and amines. Further, it is used as a reference compound for the calibration of Mossbauer spectrometer. It is also used for chromatographic detection of peptides containing cystine. It finds application as a color reagent in the development of thin layer chromatography in combination with acetaldehyde. It is also used in presence of buffers as reagent for ketone strips, which is utilized to test ketone in the urine of a diabetic.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only